Learn More
Grainger Sq. End Mill Single End Carb 1/8
Catalog No. 502749529
Encompass_Preferred
The Micro 100 Square End Mill Carbide 14T310 is a square end milling cutter made from solid carbide with a 14mm cutting diameter and 3mm corner radius. It has 4 flutes and is designed for slotting, side milling and contouring operations in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. This end mill features a TiAlN coating for increased tool life and heat resistance. It has a 7/8 inch shank diameter and comes with a total overall length of 3 inches. The end mill's square shape allows milling of slots with sharp corners.
Catalog No. 50-274-9529 Supplier Grainger Supplier No. 14T310
The Micro 100 Square End Mill Carbide 14T310 is a square end milling cutter made from solid carbide with a 14mm cutting diameter and 3mm corner radius. It has 4 flutes and is designed for slotting, side milling and contouring operations in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. This end mill features a TiAlN coating for increased tool life and heat resistance. It has a 7/8 inch shank diameter and comes with a total overall length of 3 inches. The end mill's square shape allows milling of slots with sharp corners.