Learn More
Grainger Sq. End Mill Single End Carb 1/4
Catalog No. 502750073
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, a 1-1/2" overall length, and a 1/2" cutter diameter. The end mill has 4 flutes and a right-hand spiral. It is uncoated and made in the USA. The end mill can be used on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass. It has an operating speed of 55-575 SFM and feed rates from 0.002-0.012 IPR.
Catalog No. 50-275-0073 Supplier Grainger Supplier No. 14M423
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, a 1-1/2" overall length, and a 1/2" cutter diameter. The end mill has 4 flutes and a right-hand spiral. It is uncoated and made in the USA. The end mill can be used on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass. It has an operating speed of 55-575 SFM and feed rates from 0.002-0.012 IPR.