Learn More
Grainger Sq. End Mill Single End Carb 1/16
Catalog No. 502750226
Encompass_Preferred
The Grainger 14M352 is a Westward brand 1/4-inch drive socket set. It contains 17 pieces including 10 sockets ranging from 4mm to 14mm, 5 hex keys, a ratchet, and an extension bar. The chrome vanadium steel sockets have a polished finish and feature laser etched size markings. This socket set comes packaged in a molded storage case for convenient portability and organization. With a lifetime replacement guarantee, the Westward 14M352 socket set provides a versatile assortment of tools for automotive and machinery repairs.
Catalog No. 50-275-0226 Supplier Grainger Supplier No. 14M352
The Grainger 14M352 is a Westward brand 1/4-inch drive socket set. It contains 17 pieces including 10 sockets ranging from 4mm to 14mm, 5 hex keys, a ratchet, and an extension bar. The chrome vanadium steel sockets have a polished finish and feature laser etched size markings. This socket set comes packaged in a molded storage case for convenient portability and organization. With a lifetime replacement guarantee, the Westward 14M352 socket set provides a versatile assortment of tools for automotive and machinery repairs.