Learn More
Grainger Sq. End Mill Single End Carb 1/8
Catalog No. 502751944
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches with a 0.03 inch corner radius. It can machine steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. The end mill has an American National Standard Stub length of cut and is made in the USA.
Catalog No. 50-275-1944 Supplier Grainger Supplier No. 14M377
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches with a 0.03 inch corner radius. It can machine steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. The end mill has an American National Standard Stub length of cut and is made in the USA.