Learn More
Grainger Sq. End Mill Single End Carb 1/16
Catalog No. 502752050
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14T284 is a square end milling cutter with a 1/4" shank diameter, 1/2" cutting diameter, 2" overall length, and 2 flute count. It is made of solid carbide and has a TiAlN coating for increased tool life and heat resistance. The square end mill features a center cutting design allowing plunge cuts and drilling operations. Recommended speeds are 20,000-50,000 RPM with feed rates from 0.001-0.004 IPR based on material being machined. It can be used for slotting, side milling and facing on materials like aluminum, steel, stainless steel, cast iron, and titanium.
Catalog No. 50-275-2050 Supplier Grainger Supplier No. 14T284
The Micro 100 Square End Mill Center Cutting 14T284 is a square end milling cutter with a 1/4" shank diameter, 1/2" cutting diameter, 2" overall length, and 2 flute count. It is made of solid carbide and has a TiAlN coating for increased tool life and heat resistance. The square end mill features a center cutting design allowing plunge cuts and drilling operations. Recommended speeds are 20,000-50,000 RPM with feed rates from 0.001-0.004 IPR based on material being machined. It can be used for slotting, side milling and facing on materials like aluminum, steel, stainless steel, cast iron, and titanium.