Learn More
Grainger Sq. End Mill Single End Carb 12.00mm
Catalog No. 502752839
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14G096 is a square end mill with a center cutting design. It is made of solid carbide and has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. This end mill features four flutes and can be run at speeds up to 25,000 RPM. It is well-suited for slotting, side milling and contouring in steel, aluminum and other non-ferrous materials. The center cutting design allows plunge cuts and drilling. Made in the USA by Micro 100, this is a high quality carbide end mill for moderate duty milling applications.
Catalog No. 50-275-2839 Supplier Grainger Supplier No. 14G096
The Micro 100 Square End Mill Center Cutting 14G096 is a square end mill with a center cutting design. It is made of solid carbide and has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. This end mill features four flutes and can be run at speeds up to 25,000 RPM. It is well-suited for slotting, side milling and contouring in steel, aluminum and other non-ferrous materials. The center cutting design allows plunge cuts and drilling. Made in the USA by Micro 100, this is a high quality carbide end mill for moderate duty milling applications.