Learn More
Grainger Sq. End Mill Single End Carb 11/64
Catalog No. 502753272
Encompass_Preferred
The 14M395 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 5 lbs of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It uses a non-conductive, non-corrosive agent that leaves no residue and is safe for use around sensitive electronics. The extinguisher has a chrome-plated steel cylinder with an enameled aluminum valve body and discharge hose. It comes equipped with a wall hook and mounting bracket for easy installation. This compact and lightweight fire extinguisher provides fast and effective fire suppression.
Catalog No. 50-275-3272 Supplier Grainger Supplier No. 14M395
The 14M395 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 5 lbs of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It uses a non-conductive, non-corrosive agent that leaves no residue and is safe for use around sensitive electronics. The extinguisher has a chrome-plated steel cylinder with an enameled aluminum valve body and discharge hose. It comes equipped with a wall hook and mounting bracket for easy installation. This compact and lightweight fire extinguisher provides fast and effective fire suppression.