Learn More
Grainger Sq. End Mill Single End Carb 1/8
Catalog No. 502749553
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14T311 is a square end milling cutter made by Micro 100. It has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. The end mill features a TiAlN coating for increased tool life and heat resistance. It has 4 flutes and can be run at speeds up to 25,000 RPM. This end mill is center cutting and designed for slotting, side milling and contouring in steels, tool steels, stainless steels and cast iron. It provides accurate 90 degree shoulder milling.
Catalog No. 50-274-9553 Supplier Grainger Supplier No. 14T311
The Micro 100 Square End Mill Center Cutting 14T311 is a square end milling cutter made by Micro 100. It has a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. The end mill features a TiAlN coating for increased tool life and heat resistance. It has 4 flutes and can be run at speeds up to 25,000 RPM. This end mill is center cutting and designed for slotting, side milling and contouring in steels, tool steels, stainless steels and cast iron. It provides accurate 90 degree shoulder milling.