Learn More
Grainger Sq. End Mill Single End Carb 1.30mm
Supplier:  Grainger 14K559
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1" overall length, and a 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastics. The end mill provides accurate cuts and smooth finishes at speeds up to 2000 SFM. It is made in the USA.
Catalog No. 50-274-9688