Learn More
Grainger Sq. End Mill Single End Carb 1/2
Catalog No. 502750035
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14G065 is a square end milling cutter made by Micro 100. It has a 1/4 inch shank diameter, 1/2 inch cutter diameter, and 1 inch overall length. The end mill features a TiAlN coating for increased tool life and heat resistance. It is designed for center cutting in low to medium carbon steels at speeds up to 6,000 RPM with .002-.005 inch per tooth feed rates. This is a high quality square end mill suitable for a variety of milling operations.
Catalog No. 50-275-0035 Supplier Grainger Supplier No. 14M268
The Micro 100 Square End Mill Center Cutting 14G065 is a square end milling cutter made by Micro 100. It has a 1/4 inch shank diameter, 1/2 inch cutter diameter, and 1 inch overall length. The end mill features a TiAlN coating for increased tool life and heat resistance. It is designed for center cutting in low to medium carbon steels at speeds up to 6,000 RPM with .002-.005 inch per tooth feed rates. This is a high quality square end mill suitable for a variety of milling operations.