Learn More
Grainger Sq. End Mill Single End Carb 11/64
Catalog No. 502753590
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a straight shank and is uncoated with an overall length of 2-1/2", shank diameter of 1/2", and flute length of 1-1/4". This end mill provides accurate cuts in steel, aluminum, and other non-ferrous materials. The square end allows plunging into workpieces and machining of pockets with square corners and edges. It is designed for use on CNC or manual milling machines.
Catalog No. 50-275-3590 Supplier Grainger Supplier No. 14F958
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a straight shank and is uncoated with an overall length of 2-1/2", shank diameter of 1/2", and flute length of 1-1/4". This end mill provides accurate cuts in steel, aluminum, and other non-ferrous materials. The square end allows plunging into workpieces and machining of pockets with square corners and edges. It is designed for use on CNC or manual milling machines.