Learn More
Grainger Sq. End Mill Single End Carb 1/2
Catalog No. 502751368
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. This 1/4" diameter, 4 flute end mill has a 1/2" LOC and is made of solid carbide for durability and long tool life. It features a center cutting design allowing plunge cuts and drilling operations. With superior edge strength and wear resistance, this end mill is well-suited for slotting, side milling and facing in low and high temp alloys. It provides excellent chip control when machining aluminum, steel, stainless steel, titanium and other materials. This is a versatile end mill for high performance milling in a range of applications.
Catalog No. 50-275-1368 Supplier Grainger Supplier No. 14M517
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. This 1/4" diameter, 4 flute end mill has a 1/2" LOC and is made of solid carbide for durability and long tool life. It features a center cutting design allowing plunge cuts and drilling operations. With superior edge strength and wear resistance, this end mill is well-suited for slotting, side milling and facing in low and high temp alloys. It provides excellent chip control when machining aluminum, steel, stainless steel, titanium and other materials. This is a versatile end mill for high performance milling in a range of applications.