Learn More
Grainger Sq. End Mill Single End Carb 1/2
Catalog No. 502751369
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, shoulders, and angular surfaces. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill features 4 flutes and is made of solid carbide for wear resistance and hardness. It has a TiAIN coating to reduce friction and heat. The end mill has an included angle of 90 degrees and is designed for high speed machining in aluminum, harder non-ferrous metals, pre-hardened steels, hardened steels up to 45 HRc, and most non-metallic materials. It can be run at speeds up to 8,000 RPM with conventional high speed steel speeds and feeds.
Catalog No. 50-275-1369 Supplier Grainger Supplier No. 14M518
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, shoulders, and angular surfaces. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill features 4 flutes and is made of solid carbide for wear resistance and hardness. It has a TiAIN coating to reduce friction and heat. The end mill has an included angle of 90 degrees and is designed for high speed machining in aluminum, harder non-ferrous metals, pre-hardened steels, hardened steels up to 45 HRc, and most non-metallic materials. It can be run at speeds up to 8,000 RPM with conventional high speed steel speeds and feeds.