Learn More
Grainger Sq. End Mill Single End Carb 1/4
Catalog No. 502751644
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, a 1-1/2" overall length, and a 1/2" cutter diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, cast iron, aluminum, and other non-ferrous metals. The end mill provides accurate cuts and smooth finish.
Catalog No. 50-275-1644 Supplier Grainger Supplier No. 14M428
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, a 1-1/2" overall length, and a 1/2" cutter diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, cast iron, aluminum, and other non-ferrous metals. The end mill provides accurate cuts and smooth finish.