Learn More
Grainger Sq. End Mill Single End Carb 1/4
Catalog No. 502752119
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.25 inches. The mill diameter is 0.375 inches with a 0.125 inch depth of cut. This end mill is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass. It can be run at speeds up to 5500 SFM.
Catalog No. 50-275-2119 Supplier Grainger Supplier No. 14M424
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.25 inches. The mill diameter is 0.375 inches with a 0.125 inch depth of cut. This end mill is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass. It can be run at speeds up to 5500 SFM.