Learn More
Grainger Sq. End Mill Single End Carb 1.00mm
Supplier:  Grainger 14G051
Encompass_Preferred
The Micro 100 Square End Mill Carbide is a high-performance end mill for milling a variety of materials. It features a square end design for milling 90 degree shoulders and slots. The end mill has 4 flutes and is made of solid carbide with TiAlN coating for wear resistance and heat resistance. It has a 1/2" shank diameter and is available in multiple diameters from 1/8" to 1/2". The square end mill is ideal for slotting, side milling, and facing on steels, cast iron, aluminum, and other non-ferrous metals. It provides an excellent finish and long tool life. The end mill is manufactured in the USA by Micro 100 Tool, a leading brand known for precision cutting tools.
Catalog No. 50-275-2278