Learn More
Grainger Sq. End Mill Single End Carb 25/64
Catalog No. 502751367
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, shoulders, and angles. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill features 4 flutes and is made of solid carbide for wear resistance and durability. It has a TiAIN coating to reduce friction and heat. The end mill provides accurate cuts in steel, cast iron, aluminum, and other non-ferrous metals. It is designed for high speed machining in both clockwise and counterclockwise directions.
Catalog No. 50-275-1367 Supplier Grainger Supplier No. 14M479
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, shoulders, and angles. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill features 4 flutes and is made of solid carbide for wear resistance and durability. It has a TiAIN coating to reduce friction and heat. The end mill provides accurate cuts in steel, cast iron, aluminum, and other non-ferrous metals. It is designed for high speed machining in both clockwise and counterclockwise directions.