Learn More
Grainger Sq. End Mill Single End Carb 2.00mm
Catalog No. 502752279
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and a 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, cast iron, aluminum, and other non-ferrous metals at speeds up to 2000 SFM. Made in the USA, this end mill provides accurate cuts and smooth milling action.
Catalog No. 50-275-2279 Supplier Grainger Supplier No. 14G057
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and a 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, cast iron, aluminum, and other non-ferrous metals at speeds up to 2000 SFM. Made in the USA, this end mill provides accurate cuts and smooth milling action.