Learn More
Grainger Sq. End Mill Single End Carb 2.00mm
Catalog No. 502749735
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel square end milling cutter with 4 flutes and a 1/2" shank diameter. It has an overall length of 3-1/2" and a cutting diameter of 1/2". The square end mill features a TiN coating for increased tool life and lubricity. It can be used for slotting, side milling and straddle milling in low carbon steels at surface speeds up to 135 SFM when taking light cuts. This durable end mill is ideal for general purpose milling in machine shops.
Catalog No. 50-274-9735 Supplier Grainger Supplier No. 14K573
The Grainger Approved Square End Mill Single End is a high-speed steel square end milling cutter with 4 flutes and a 1/2" shank diameter. It has an overall length of 3-1/2" and a cutting diameter of 1/2". The square end mill features a TiN coating for increased tool life and lubricity. It can be used for slotting, side milling and straddle milling in low carbon steels at surface speeds up to 135 SFM when taking light cuts. This durable end mill is ideal for general purpose milling in machine shops.