Learn More
Grainger Sq. End Mill Single End Carb 3/8
Catalog No. 502748825
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is uncoated and made in the USA. The end mill can be used on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass at speeds up to 2000 SFM.
Catalog No. 50-274-8825 Supplier Grainger Supplier No. 14M682
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1-1/2" overall length, and 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is uncoated and made in the USA. The end mill can be used on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass at speeds up to 2000 SFM.