Learn More
Grainger Sq. End Mill Single End Carb 3/16
Catalog No. 502748836
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14F965 is a square end milling tool made by Micro 100. It has a 1/4 inch shank diameter, 1/2 inch cutting diameter, and 2 inch overall length. The end mill is center cutting and has 4 flutes. It is made of solid carbide and can machine steels, cast iron, aluminum, copper, brass, and plastics. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for high speed milling in light to medium duty applications. The end mill provides accurate 90 degree shoulders in slotting, side milling, and facing operations.
Catalog No. 50-274-8836 Supplier Grainger Supplier No. 14F965
The Micro 100 Square End Mill Center Cutting 14F965 is a square end milling tool made by Micro 100. It has a 1/4 inch shank diameter, 1/2 inch cutting diameter, and 2 inch overall length. The end mill is center cutting and has 4 flutes. It is made of solid carbide and can machine steels, cast iron, aluminum, copper, brass, and plastics. The end mill has a TiAlN coating for increased tool life and heat resistance. It is designed for high speed milling in light to medium duty applications. The end mill provides accurate 90 degree shoulders in slotting, side milling, and facing operations.