Learn More
Grainger Sq. End Mill Single End Carb 3/16
Catalog No. 502749289
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14F961 is a square end milling cutter with a center cutting edge. It has a 1/4" shank diameter and a 1/2" cutting diameter. The overall length is 2-1/2". It is made of solid carbide for wear resistance and has 2 flutes. The square end allows milling square slots and contours. The center cutting edge allows plunging into the workpiece. It can be run at speeds up to 325 SFM. This is a versatile square end mill for a variety of milling operations.
Catalog No. 50-274-9289 Supplier Grainger Supplier No. 14F961
The Micro 100 Square End Mill Center Cutting 14F961 is a square end milling cutter with a center cutting edge. It has a 1/4" shank diameter and a 1/2" cutting diameter. The overall length is 2-1/2". It is made of solid carbide for wear resistance and has 2 flutes. The square end allows milling square slots and contours. The center cutting edge allows plunging into the workpiece. It can be run at speeds up to 325 SFM. This is a versatile square end mill for a variety of milling operations.