Learn More
Grainger Sq. End Mill Single End Carb 3.00mm
Catalog No. 502753086
Encompass_Preferred
The Grainger 14G062 is a Westward brand 1/4-inch drive socket set that includes both SAE and metric sockets. The set contains 21 sockets ranging in size from 4mm to 15mm and 1/8 inch to 9/16 inch. The chrome vanadium sockets have a matte finish and feature a knurled design for easy hand tightening. The set comes organized in a molded plastic case for convenient storage and portability. With both common and hard-to-find sizes, this socket set is great for automotive and machinery repair and maintenance.
Catalog No. 50-275-3086 Supplier Grainger Supplier No. 14G062
The Grainger 14G062 is a Westward brand 1/4-inch drive socket set that includes both SAE and metric sockets. The set contains 21 sockets ranging in size from 4mm to 15mm and 1/8 inch to 9/16 inch. The chrome vanadium sockets have a matte finish and feature a knurled design for easy hand tightening. The set comes organized in a molded plastic case for convenient storage and portability. With both common and hard-to-find sizes, this socket set is great for automotive and machinery repair and maintenance.