Learn More
Grainger Sq. End Mill Single End Carb 3/8
Catalog No. 502751380
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide 3/8" is a square end milling cutter with a 3/8" shank diameter and a 1/2" cutting diameter. It has 4 flutes and is made of solid carbide for wear resistance and durability. This end mill is designed for slotting, side milling and contour milling in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. It has a TiAlN coating for heat and wear resistance. The overall length is 3" and it has a 1/2" LOC. This is a high quality carbide end mill made in the USA.
Catalog No. 50-275-1380 Supplier Grainger Supplier No. 14T172
The Grainger Approved Square End Mill Single End Carbide 3/8" is a square end milling cutter with a 3/8" shank diameter and a 1/2" cutting diameter. It has 4 flutes and is made of solid carbide for wear resistance and durability. This end mill is designed for slotting, side milling and contour milling in steel, stainless steel, cast iron, aluminum and other non-ferrous metals. It has a TiAlN coating for heat and wear resistance. The overall length is 3" and it has a 1/2" LOC. This is a high quality carbide end mill made in the USA.