Learn More
Grainger Sq. End Mill Single End Carb 3/8
Catalog No. 502751686
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank diameter. It has an American National standard taper shank and is made in the USA. The end mill's composition is M42 high-speed steel which provides good wear resistance and allows higher cutting speeds than regular high-speed steel. It can be used on steel, cast iron, aluminum, and other non-ferrous metals.
Catalog No. 50-275-1686 Supplier Grainger Supplier No. 14M473
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank diameter. It has an American National standard taper shank and is made in the USA. The end mill's composition is M42 high-speed steel which provides good wear resistance and allows higher cutting speeds than regular high-speed steel. It can be used on steel, cast iron, aluminum, and other non-ferrous metals.