Learn More
Grainger Sq. End Mill Single End Carb 3/8
Catalog No. 502751687
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.25 inches. This end mill is uncoated and made in the USA. The maximum RPM is 2700. It can be used on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastics.
Catalog No. 50-275-1687 Supplier Grainger Supplier No. 14M476
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.25 inches. This end mill is uncoated and made in the USA. The maximum RPM is 2700. It can be used on steel, stainless steel, cast iron, aluminum, brass, bronze, and plastics.