Learn More
Grainger Sq. End Mill Single End Carb 3.00mm
Catalog No. 502751931
Encompass_Preferred
The Grainger 14G061 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and delay, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards. This auto-darkening welding helmet is designed for reliability and durability.
Catalog No. 50-275-1931 Supplier Grainger Supplier No. 14G061
The Grainger 14G061 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and delay, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards. This auto-darkening welding helmet is designed for reliability and durability.