Learn More
Grainger Sq. End Mill Single End Carb 3/16
Catalog No. 502753330
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. It features a square end design with center cutting capability for plunging, slotting and pocketing operations. The end mill has four flutes and is made of solid carbide for durability and tool life. It has a 1/4 inch shank diameter and is available in multiple diameters from 1/8 inch to 1/2 inch for different milling requirements. The center cutting design allows milling by plunging as well as conventional milling. Overall, this is a versatile square end mill suitable for high-performance milling in a range of materials.
Catalog No. 50-275-3330 Supplier Grainger Supplier No. 14M404
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. It features a square end design with center cutting capability for plunging, slotting and pocketing operations. The end mill has four flutes and is made of solid carbide for durability and tool life. It has a 1/4 inch shank diameter and is available in multiple diameters from 1/8 inch to 1/2 inch for different milling requirements. The center cutting design allows milling by plunging as well as conventional milling. Overall, this is a versatile square end mill suitable for high-performance milling in a range of materials.