Learn More
Grainger Sq. End Mill Single End Carb 3/64
Catalog No. 502753354
Encompass_Preferred
The Micro 100 Square End Mill Carbide is a square end milling cutter made from solid carbide with a TiAlN coating for wear resistance. It has 4 flutes and a 1/2" shank diameter. The overall length is 3" and it has a 1/2" cutting diameter with square corners. This is designed for slotting, side milling and contour milling in steels, tool steels, cast iron and other materials. It provides accurate 90 degree shoulders. The TiAlN coating helps resist welding and buildup on the cutter.
Catalog No. 50-275-3354 Supplier Grainger Supplier No. 14T282
The Micro 100 Square End Mill Carbide is a square end milling cutter made from solid carbide with a TiAlN coating for wear resistance. It has 4 flutes and a 1/2" shank diameter. The overall length is 3" and it has a 1/2" cutting diameter with square corners. This is designed for slotting, side milling and contour milling in steels, tool steels, cast iron and other materials. It provides accurate 90 degree shoulders. The TiAlN coating helps resist welding and buildup on the cutter.