Learn More
Grainger Sq. End Mill Single End Carb 3/32
Catalog No. 502748859
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide 3/32" is a square end milling cutter with a 3/32" cutting diameter. It has a 1/4" shank diameter and an overall length of 2-1/2". The end mill features four flutes and is made of solid carbide for wear resistance and durability. This single end mill is designed for light milling in low horsepower machines. It can be used for slotting, side milling and profiling in materials like aluminum, brass, bronze, plastics and woods.
Catalog No. 50-274-8859 Supplier Grainger Supplier No. 14T297
The Grainger Approved Square End Mill Single End Carbide 3/32" is a square end milling cutter with a 3/32" cutting diameter. It has a 1/4" shank diameter and an overall length of 2-1/2". The end mill features four flutes and is made of solid carbide for wear resistance and durability. This single end mill is designed for light milling in low horsepower machines. It can be used for slotting, side milling and profiling in materials like aluminum, brass, bronze, plastics and woods.