Learn More
Grainger Sq. End Mill Single End Carb 4.00mm
Catalog No. 502751549
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill for milling a variety of materials. It features a square end design for milling 90-degree shoulders and angles. The center cutting design allows plunge cuts into a workpiece. It is made of solid carbide for durability and has 2 flutes for efficient chip removal. The end mill has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2" for different milling applications. Overall, this is a versatile square end mill suitable for shoulder milling, slotting, side milling, and contouring operations.
Catalog No. 50-275-1549 Supplier Grainger Supplier No. 14G065
The Micro 100 Square End Mill Center Cutting is a high-performance end mill for milling a variety of materials. It features a square end design for milling 90-degree shoulders and angles. The center cutting design allows plunge cuts into a workpiece. It is made of solid carbide for durability and has 2 flutes for efficient chip removal. The end mill has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2" for different milling applications. Overall, this is a versatile square end mill suitable for shoulder milling, slotting, side milling, and contouring operations.