Learn More
Grainger Sq. End Mill Single End Carb 4.00mm
Catalog No. 502752675
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand cut. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank neck length. It has an American National standard taper shank and is made in the USA. The end mill's composition is M2 high speed steel which provides good wear resistance and allows higher cutting speeds than carbon steel end mills. It can machine steels, cast iron, aluminum, brass and bronze. This is a good general purpose end mill for a variety of materials.
Catalog No. 50-275-2675 Supplier Grainger Supplier No. 14G069
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has 4 flutes and a right-hand cut. The overall length is 3 inches and the shank diameter is 1/2 inch. The mill diameter is 1 inch with a 1/8 inch shank neck length. It has an American National standard taper shank and is made in the USA. The end mill's composition is M2 high speed steel which provides good wear resistance and allows higher cutting speeds than carbon steel end mills. It can machine steels, cast iron, aluminum, brass and bronze. This is a good general purpose end mill for a variety of materials.