Learn More
Grainger Sq. End Mill Single End Carb 5/32
Catalog No. 502749324
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14F944 is a square end milling cutter made by Micro 100. It has a 1/4 inch shank diameter, 1/2 inch cutter diameter, 2 inch overall length, and 2 flute count. This carbide end mill is center cutting and has square ends for milling 90 degree shoulders. It can machine steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. With its sharp cutting edges, this end mill provides accurate right angle shoulders in slotting, side milling, and facing operations.
Catalog No. 50-274-9324 Supplier Grainger Supplier No. 14F949
The Micro 100 Square End Mill Center Cutting 14F944 is a square end milling cutter made by Micro 100. It has a 1/4 inch shank diameter, 1/2 inch cutter diameter, 2 inch overall length, and 2 flute count. This carbide end mill is center cutting and has square ends for milling 90 degree shoulders. It can machine steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. With its sharp cutting edges, this end mill provides accurate right angle shoulders in slotting, side milling, and facing operations.