Learn More
Grainger Sq. End Mill Single End Carb 5/32
Catalog No. 502749179
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. This 1/4" diameter, 2 flute end mill has a square end for milling 90 degree shoulders and slots in aluminum, steel, stainless steel, cast iron, titanium, plastic and other materials. It features a TiAlN coating for heat and wear resistance during high speed machining. The center cutting design allows plunge cutting and drilling. Recommended speed is 25,000 RPM with a feed rate of 10 IPM. It has a shank diameter of 1/4" and an overall length of 2-1/2". Made in the USA by Micro 100 Tool Corp.
Catalog No. 50-274-9179 Supplier Grainger Supplier No. 14F950
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a wide range of materials. This 1/4" diameter, 2 flute end mill has a square end for milling 90 degree shoulders and slots in aluminum, steel, stainless steel, cast iron, titanium, plastic and other materials. It features a TiAlN coating for heat and wear resistance during high speed machining. The center cutting design allows plunge cutting and drilling. Recommended speed is 25,000 RPM with a feed rate of 10 IPM. It has a shank diameter of 1/4" and an overall length of 2-1/2". Made in the USA by Micro 100 Tool Corp.