Learn More
Grainger Sq. End Mill Single End Carb 5/32
Catalog No. 502748778
Encompass_Preferred
The Grainger Sq End Mill Single End Carbide 5/32" is a square end milling cutter made from solid carbide with a 5/32" cutting diameter. It has a 1/4" shank diameter and is uncoated with an overall length of 2-1/2". This single end mill is designed for slotting, side milling and contour milling in steel, cast iron, aluminum and other non-ferrous metals. It provides good chip flow in milling applications. The carbide construction gives a long tool life with faster speeds and feeds compared to high speed steel.
Catalog No. 50-274-8778 Supplier Grainger Supplier No. 14F948
The Grainger Sq End Mill Single End Carbide 5/32" is a square end milling cutter made from solid carbide with a 5/32" cutting diameter. It has a 1/4" shank diameter and is uncoated with an overall length of 2-1/2". This single end mill is designed for slotting, side milling and contour milling in steel, cast iron, aluminum and other non-ferrous metals. It provides good chip flow in milling applications. The carbide construction gives a long tool life with faster speeds and feeds compared to high speed steel.