Learn More
Grainger Sq. End Mill Single End Carb 5/32
Catalog No. 502748841
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, keyways, pockets, and other 90 degree surfaces. It has a 1/2 inch shank diameter, 1 inch cutting diameter, 2 inch overall length, and 2 flutes. The end mill is made of solid carbide for wear resistance and has TiAlN coating for heat resistance and lubricity. It can machine steels, cast iron, aluminum, and other non-ferrous metals at high speeds and feeds. This is a high quality carbide end mill suitable for roughing and finishing operations.
Catalog No. 50-274-8841 Supplier Grainger Supplier No. 14F953
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining slots, keyways, pockets, and other 90 degree surfaces. It has a 1/2 inch shank diameter, 1 inch cutting diameter, 2 inch overall length, and 2 flutes. The end mill is made of solid carbide for wear resistance and has TiAlN coating for heat resistance and lubricity. It can machine steels, cast iron, aluminum, and other non-ferrous metals at high speeds and feeds. This is a high quality carbide end mill suitable for roughing and finishing operations.