Learn More
Grainger Sq. End Mill Single End Carb 5/64
Catalog No. 502750010
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. This end mill is uncoated and made in the USA. The maximum RPM is 2700. It can slot to a depth of 1.5 inches in steel and 2 inches in aluminum. This is a good general purpose end mill for light milling applications.
Catalog No. 50-275-0010 Supplier Grainger Supplier No. 14M356
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. This end mill is uncoated and made in the USA. The maximum RPM is 2700. It can slot to a depth of 1.5 inches in steel and 2 inches in aluminum. This is a good general purpose end mill for light milling applications.