Learn More
Grainger Sq. End Mill Single End Carb 5/64
Catalog No. 502750039
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches. This end mill is uncoated and made in the USA. It can be used for slotting, side milling, and facing on low carbon steels at surface speeds up to 120 SFM when paired with rigid setups.
Catalog No. 50-275-0039 Supplier Grainger Supplier No. 14M360
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches. This end mill is uncoated and made in the USA. It can be used for slotting, side milling, and facing on low carbon steels at surface speeds up to 120 SFM when paired with rigid setups.