Learn More
Grainger Sq. End Mill Single End Carb 5/32
Catalog No. 502750456
Encompass_Preferred
The 14M386 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", Very Fine grade. It features an open web construction and is made of high-quality fibers for cleaning, scouring, and finishing applications. This abrasive hand pad has a flexible backing that conforms to surfaces. It effectively removes rust, oxidation, and light scratches without harming the base material. The very fine grade pad is ideal for light scrubbing and finishing work. It can be used wet or dry on metal, plastic, glass and other surfaces. This durable hand pad measures 6 inches by 9 inches.
Catalog No. 50-275-0456 Supplier Grainger Supplier No. 14M386
The 14M386 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", Very Fine grade. It features an open web construction and is made of high-quality fibers for cleaning, scouring, and finishing applications. This abrasive hand pad has a flexible backing that conforms to surfaces. It effectively removes rust, oxidation, and light scratches without harming the base material. The very fine grade pad is ideal for light scrubbing and finishing work. It can be used wet or dry on metal, plastic, glass and other surfaces. This durable hand pad measures 6 inches by 9 inches.