Learn More
Grainger Sq. End Mill Single End Carb 5/32
Catalog No. 502751946
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2 inch shank diameter, 1 inch overall length, and a 1/2 inch cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass at speeds up to 2000 SFM. This end mill is made in the USA.
Catalog No. 50-275-1946 Supplier Grainger Supplier No. 14M388
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2 inch shank diameter, 1 inch overall length, and a 1/2 inch cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and fiberglass at speeds up to 2000 SFM. This end mill is made in the USA.