Learn More
Grainger Sq. End Mill Single End Carb 5/16
Catalog No. 502749404
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches with a 0.03 inch corner radius. It can machine aluminum, brass, bronze, cast iron, steel, and most alloys. This end mill is designed for high feed rates and productivity in light milling applications.
Catalog No. 50-274-9404 Supplier Grainger Supplier No. 14M713
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.5 inches. The mill diameter is 0.625 inches with a 0.03 inch corner radius. It can machine aluminum, brass, bronze, cast iron, steel, and most alloys. This end mill is designed for high feed rates and productivity in light milling applications.