Learn More
Grainger Sq. End Mill Single End Carb 5/64
Catalog No. 502748677
Encompass_Preferred
The Micro 100 Square End Mill Carbide is a 14T290 milling cutter with a square end for machining slots, keyways, and grooves. It has a 1/2" shank diameter and an overall length of 2-1/2". The carbide cutter has 4 flutes and a right-hand spiral. It can machine steel, cast iron, aluminum, and other non-ferrous metals. The square end mill has TiAlN coating for heat and wear resistance. Recommended speed is 60-120 SFM with a 0.002-0.010" chip load per tooth.
Catalog No. 50-274-8677 Supplier Grainger Supplier No. 14T290
The Micro 100 Square End Mill Carbide is a 14T290 milling cutter with a square end for machining slots, keyways, and grooves. It has a 1/2" shank diameter and an overall length of 2-1/2". The carbide cutter has 4 flutes and a right-hand spiral. It can machine steel, cast iron, aluminum, and other non-ferrous metals. The square end mill has TiAlN coating for heat and wear resistance. Recommended speed is 60-120 SFM with a 0.002-0.010" chip load per tooth.