Learn More
Grainger Sq. End Mill Single End Carb 7/64
Catalog No. 502751468
Encompass_Preferred
The 14M373 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", Very Fine grade. It features an open textured web construction and is made of high-quality fibers for cleaning, scouring, and finishing on surfaces. This hand pad has durable fibers that resist shredding and matting. It can be used for light scrubbing and finishing work on metal, plastic, glass, ceramic, rubber, and painted wood surfaces. The very fine grade makes it ideal for applying final finishes. This hand pad measures 6 inches by 9 inches.
Catalog No. 50-275-1468 Supplier Grainger Supplier No. 14M373
The 14M373 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", Very Fine grade. It features an open textured web construction and is made of high-quality fibers for cleaning, scouring, and finishing on surfaces. This hand pad has durable fibers that resist shredding and matting. It can be used for light scrubbing and finishing work on metal, plastic, glass, ceramic, rubber, and painted wood surfaces. The very fine grade makes it ideal for applying final finishes. This hand pad measures 6 inches by 9 inches.