Learn More
Grainger Sq. End Mill Single End Carb 9/64
Catalog No. 502749178
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14F944 is a square end milling cutter with a center cutting edge. It has a 1/4 inch shank diameter and a 1/2 inch cutting diameter. The overall length is 2 inches. This carbide end mill is designed for slotting, side milling and contour milling in steel, cast iron, aluminum and other non-ferrous metals. It features four flutes and a right-hand spiral. The center cutting edge allows plunge cuts. Made in the USA by Micro 100 Tool Corp.
Catalog No. 50-274-9178 Supplier Grainger Supplier No. 14F944
The Micro 100 Square End Mill Center Cutting 14F944 is a square end milling cutter with a center cutting edge. It has a 1/4 inch shank diameter and a 1/2 inch cutting diameter. The overall length is 2 inches. This carbide end mill is designed for slotting, side milling and contour milling in steel, cast iron, aluminum and other non-ferrous metals. It features four flutes and a right-hand spiral. The center cutting edge allows plunge cuts. Made in the USA by Micro 100 Tool Corp.