Learn More
Grainger Sq. End Mill Single End Carb 9/32
Catalog No. 502752063
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1" overall length, and a 1/4" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, cast iron, aluminum, and other non-ferrous metals at speeds up to 2000 SFM. The end mill is made in the USA by Grainger Approved to tight tolerances for accurate cuts.
Catalog No. 50-275-2063 Supplier Grainger Supplier No. 14M439
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has a 1/2" shank diameter, 1" overall length, and a 1/4" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is designed for use on steel, cast iron, aluminum, and other non-ferrous metals at speeds up to 2000 SFM. The end mill is made in the USA by Grainger Approved to tight tolerances for accurate cuts.