Learn More
Grainger Sq. End Mill Single End Carb 9/64
Supplier:  Grainger 14F943
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14F943 is a square end milling cutter made by Micro 100. It has a 1/4 inch shank diameter, 1/2 inch cutting diameter, 2 inch overall length, and 2 flute design. The end mill is center cutting and made of solid carbide for hardness and wear resistance. It can be used for slotting, side milling and contouring operations in steel, aluminum and other materials. The square shape allows milling of 90 degree shoulders and corners.
Catalog No. 50-275-3618