Learn More
Grainger Sq. End Mill Single End Carb 9/64
Catalog No. 502751706
Encompass_Preferred
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches with a 0.03 inch corner radius. It can machine steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. It has an American National Standard Stub length shank. The end mill is uncoated.
Catalog No. 50-275-1706 Supplier Grainger Supplier No. 14M385
The Grainger Approved Square End Mill Single End is a high-speed steel end mill with a square end for milling slots, keyways, and pockets. It has four flutes and a right-hand spiral. The overall length is 3 inches and the shank diameter is 0.375 inches. The mill diameter is 0.5 inches with a 0.03 inch corner radius. It can machine steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. It has an American National Standard Stub length shank. The end mill is uncoated.