Learn More
Grainger Sq. End Mill Single End Carb 9/64
Catalog No. 502748777
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting 14F946 is a high-performance end mill for milling a variety of materials. This carbide end mill features a square end design for milling 90 degree shoulders and angles. With a diameter of 1/4", this end mill has a center cutting design that allows plunge cuts into the middle of a workpiece. The 14F946 has a 2 flute design and is uncoated for general purpose milling applications. Made in the USA, this end mill is ideal for use in CNC machining centers for milling slots, shoulders, and contours in steels, aluminum, and other materials.
Catalog No. 50-274-8777 Supplier Grainger Supplier No. 14F946
The Micro 100 Square End Mill Center Cutting 14F946 is a high-performance end mill for milling a variety of materials. This carbide end mill features a square end design for milling 90 degree shoulders and angles. With a diameter of 1/4", this end mill has a center cutting design that allows plunge cuts into the middle of a workpiece. The 14F946 has a 2 flute design and is uncoated for general purpose milling applications. Made in the USA, this end mill is ideal for use in CNC machining centers for milling slots, shoulders, and contours in steels, aluminum, and other materials.