Learn More
Grainger Sq. End Mill Single End Carb 9/64
Catalog No. 502748843
Encompass_Preferred
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a variety of materials. This 1/4" diameter, 2 flute end mill features a square end for milling 90 degree shoulders and slots in aluminum, steel, stainless steel, cast iron, titanium, plastic and other materials. The center cutting design allows plunge cuts into the middle of a workpiece. The tool is made from solid carbide for durability and features TiAlN coating for heat resistance and lubricity. Recommended speed is 20,000 RPM with feed rates from 0.001 to 0.004 IPR based on material.
Catalog No. 50-274-8843 Supplier Grainger Supplier No. 14F945
The Micro 100 Square End Mill Center Cutting is a high-performance end mill designed for milling a variety of materials. This 1/4" diameter, 2 flute end mill features a square end for milling 90 degree shoulders and slots in aluminum, steel, stainless steel, cast iron, titanium, plastic and other materials. The center cutting design allows plunge cuts into the middle of a workpiece. The tool is made from solid carbide for durability and features TiAlN coating for heat resistance and lubricity. Recommended speed is 20,000 RPM with feed rates from 0.001 to 0.004 IPR based on material.