Learn More
Grainger Sq. End Mill Single End Carb 9/64
Catalog No. 502749275
Encompass_Preferred
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining square slots, pockets, and edges. It has a 1/2" shank diameter and an overall length of 3". The carbide cutter is made of solid carbide for wear resistance and has 2 flutes. This end mill has a right hand spiral flute direction and is center cutting for plunging. It can machine steels, cast iron, aluminum, and other non-ferrous metals at high speeds. The end mill has an uncoated carbide grade for general purpose milling applications.
Catalog No. 50-274-9275 Supplier Grainger Supplier No. 14F947
The Grainger Approved Square End Mill Single End Carbide is a milling cutter with a square end for machining square slots, pockets, and edges. It has a 1/2" shank diameter and an overall length of 3". The carbide cutter is made of solid carbide for wear resistance and has 2 flutes. This end mill has a right hand spiral flute direction and is center cutting for plunging. It can machine steels, cast iron, aluminum, and other non-ferrous metals at high speeds. The end mill has an uncoated carbide grade for general purpose milling applications.